Assay Details
Target Gene Details
Entrez Gene ID: | 200895 |
Gene Name: | dihydrofolate reductase 2 |
Gene Aliases: |
DHFRL1, DHFRP4 |
Location: |
Chr.3:94057922-94063656 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHFR2 | NM_001195643.1 | NP_001182572.1 | ||
| NM_176815.4 | NP_789785.1 | |||
| XM_011512537.2 | XP_011510839.1 | |||
| AK122674.1 | ||||
| AL832912.1 | CAH10617.1 | |||
| BC045541.1 | AAH45541.1 | |||
| BC063379.1 | AAH63379.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893746 | Chr.3:93792469 - 94164796 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map