Assay Details
Target Gene Details
Entrez Gene ID: | 83850 |
Gene Name: | extended synaptotagmin 3 |
Gene Aliases: |
CHR3SYT, E-Syt3, FAM62C |
Location: |
Chr.3:138434573-138479942 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESYT3 | NM_001322831.1 | NP_001309760.1 | ||
| NM_001322834.1 | NP_001309763.1 | |||
| NM_031913.4 | NP_114119.2 | |||
| NR_136409.1 | ||||
| AK125080.1 | ||||
| AK289530.1 | ||||
| DQ993202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999041 | Chr.3:138429970 - 138629146 on Build GRCh38 | Gain |
|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
| nsv591853 | Chr.3:138381697 - 138475593 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116492611] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map