Assay Details
Target Gene Details
Entrez Gene ID: | 344758 |
Gene Name: | G protein-coupled receptor 149 |
Gene Aliases: |
IEDA, PGR10 |
Location: |
Chr.3:154337672-154429715 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR149 | NM_001038705.1 | NP_001033794.1 | ||
| AY255534.1 | AAO85046.1 | |||
| DQ318360.1 | ABC49857.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72998669] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map