Assay Details
Target Gene Details
Entrez Gene ID: | 8801 |
Gene Name: | succinate-CoA ligase GDP-forming beta subunit |
Gene Aliases: |
GBETA |
Location: |
Chr.3:67240957-67654614 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUCLG2 | XM_017007415.1 | XP_016862904.1 | ||
| XM_017007419.1 | XP_016862908.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003158 | Chr.3:66999779 - 68395539 on Build GRCh38 | Loss |
|
| esv3893718 | Chr.3:67266860 - 67294799 on Build GRCh38 | Loss |
|
| esv3596450 | Chr.3:67145428 - 67286310 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75990230] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map