Assay Details
Target Gene Details
Entrez Gene ID: | 285194 |
Gene Name: | tumor suppressor candidate 7 (non-protein coding) |
Gene Aliases: |
LINC00902, LSAMP-AS1, LSAMP-AS3, LSAMPAS3, NCRNA00295 |
Location: |
Chr.3:116709788-116717040 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUSC7 | NR_015391.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829686 | Chr.3:116606775 - 116775694 on Build GRCh38 | Gain |
|
| esv3893770 | Chr.3:116409243 - 116721198 on Build GRCh38 | Gain |
|
| dgv295n21 | Chr.3:116676731 - 116866434 on Build GRCh38 | Loss |
|
| nsv1011135 | Chr.3:116380194 - 119036472 on Build GRCh38 | Gain |
|

Genomic Map