Assay Details
Target Gene Details
Entrez Gene ID: | 100534599 |
Gene Name: | ISY1-RAB43 readthrough |
Gene Aliases: |
- |
Location: |
Chr.3:129087569-129161230 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISY1-RAB43 | NM_001204890.1 | NP_001191819.1 | ||
| AB032986.1 |
Target Gene Details
Entrez Gene ID: | 339122 |
Gene Name: | RAB43, member RAS oncogene family |
Gene Aliases: |
RAB11B, RAB41 |
Location: |
Chr.3:129087569-129122150 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB43 | NM_001204883.1 | NP_001191812.1 | ||
| NM_001204884.1 | NP_001191813.1 | |||
| NM_001204885.1 | NP_001191814.1 | |||
| NM_001204886.1 | NP_001191815.1 | |||
| NM_001204887.1 | NP_001191816.1 | |||
| NM_001204888.1 | NP_001191817.1 | |||
| NM_198490.2 | NP_940892.1 | |||
| AK128345.1 | ||||
| AK291014.1 | ||||
| AY166852.2 | AAO17291.1 | |||
| BC062319.1 | AAH62319.1 | |||
| BX331971.2 | ||||
| D45418.1 | ||||
| DA008447.1 | ||||
| DA103180.1 | ||||
| DA317502.1 | ||||
| DA339537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591650 | Chr.3:128986168 - 129177231 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map