Assay Details
Target Gene Details
Entrez Gene ID: | 378 |
Gene Name: | ADP ribosylation factor 4 |
Gene Aliases: |
ARF2 |
Location: |
Chr.3:57571363-57597488 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARF4 | NM_001660.3 | NP_001651.1 | ||
| AF493883.1 | AAM12597.1 | |||
| AK313008.1 | ||||
| BC003364.1 | AAH03364.1 | |||
| BC008753.2 | AAH08753.1 | |||
| BC016325.1 | AAH16325.1 | |||
| BC022866.1 | AAH22866.1 | |||
| M36341.1 | AAA53081.1 |
Target Gene Details
Entrez Gene ID: | 201626 |
Gene Name: | phosphodiesterase 12 |
Gene Aliases: |
2'-PDE, 2-PDE |
Location: |
Chr.3:57556247-57656480 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE12 | NM_001322176.1 | NP_001309105.1 | ||
| BM713922.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008684 | Chr.3:57537986 - 57766490 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map