Assay Details
Target Gene Details
Entrez Gene ID: | 9779 |
Gene Name: | TBC1 domain family member 5 |
Gene Aliases: |
- |
Location: |
Chr.3:17157162-17742739 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008636 | Chr.3:17131923 - 17166775 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112962032] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map