Assay Details
Target Gene Details
Entrez Gene ID: | 5922 |
Gene Name: | RAS p21 protein activator 2 |
Gene Aliases: |
GAP1M |
Location: |
Chr.3:141487047-141615363 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RASA2 | NM_001303245.1 | NP_001290174.1 | ||
NM_001303246.1 | NP_001290175.1 | |||
NM_006506.3 | NP_006497.2 | |||
AF115573.1 | AAD09821.1 | |||
AK292016.1 | ||||
AK301015.1 | ||||
D78155.1 | BAA11230.1 | |||
D82880.1 | BAA11621.1 | |||
DA792257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv955358 | Chr.3:141490759 - 141499758 on Build GRCh38 | Duplication |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
