Assay Details
Target Gene Details
Entrez Gene ID: | 1295 |
Gene Name: | collagen type VIII alpha 1 |
Gene Aliases: |
C3orf7 |
Location: |
Chr.3:99638596-99796733 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL8A1 | NM_001850.4 | NP_001841.2 | ||
| NM_020351.3 | NP_065084.2 | |||
| AF170702.1 | AAF89665.1 | |||
| AK311449.1 | ||||
| BC013581.1 | AAH13581.1 | |||
| CA393523.1 | ||||
| CN484310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597151 | Chr.3:99516655 - 99690858 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78595788] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map