Assay Details
Target Gene Details
Entrez Gene ID: | 64332 |
Gene Name: | NFKB inhibitor zeta |
Gene Aliases: |
IKBZ, INAP, MAIL |
Location: |
Chr.3:101827990-101861025 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFKBIZ | NM_001005474.2 | NP_001005474.1 | ||
| AB074898.1 | ||||
| AF548362.1 | AAN40698.1 | |||
| AK054787.1 | ||||
| BM050734.1 | ||||
| DA541852.1 | ||||
| DQ224339.1 | ABB02425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953238 | Chr.3:101825757 - 101845056 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map