Assay Details
Target Gene Details
Entrez Gene ID: | 2596 |
Gene Name: | growth associated protein 43 |
Gene Aliases: |
B-50, PP46 |
Location: |
Chr.3:115623304-115721487 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAP43 | NM_001130064.1 | NP_001123536.1 | ||
| NM_002045.3 | NP_002036.1 | |||
| AF279774.1 | ||||
| AK289699.1 | ||||
| AK290100.1 | ||||
| AK313658.1 | ||||
| BC007936.2 | AAH07936.1 | |||
| BT019771.1 | AAV38576.1 | |||
| BT019810.1 | AAV38613.1 | |||
| BX648458.1 | ||||
| DA701390.1 | ||||
| M25667.1 | AAA52516.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72961636] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map