Assay Details
Target Gene Details
Entrez Gene ID: | 57560 |
Gene Name: | intraflagellar transport 80 |
Gene Aliases: |
ATD2, SRTD2, WDR56 |
Location: |
Chr.3:160256986-160399532 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFT80 | NM_001190241.1 | NP_001177170.1 | ||
| NM_001190242.1 | NP_001177171.1 | |||
| NM_020800.2 | NP_065851.1 | |||
| AB037795.1 | ||||
| AK055904.1 | ||||
| AK125342.1 | ||||
| AK303410.1 | ||||
| AL133045.1 | CAB61372.1 | |||
| BC042027.1 | AAH42027.1 | |||
| BC101494.1 | AAI01495.1 | |||
| BC113669.1 |
More Information
Set Membership: |
|

Genomic Map