Assay Details
Target Gene Details
Entrez Gene ID: | 28990 |
Gene Name: | asteroid homolog 1 (Drosophila) |
Gene Aliases: |
HT001 |
Location: |
Chr.3:131013873-131026854 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASTE1 | NM_001288950.1 | NP_001275879.1 | ||
NM_014065.3 | NP_054784.2 | |||
NR_110229.1 | ||||
XM_017006266.1 | XP_016861755.1 | |||
XM_017006267.1 | XP_016861756.1 | |||
XM_017006268.1 | XP_016861757.1 | |||
XM_017006269.1 | XP_016861758.1 | |||
AF113539.1 | AAF14876.1 | |||
AK027258.1 | ||||
AK027806.1 | BAB55383.1 | |||
AK294156.1 | ||||
AK301388.1 | ||||
BC030274.1 | ||||
BC103709.1 | AAI03710.1 | |||
BC110616.2 | AAI10617.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
