Assay Details
Target Gene Details
Entrez Gene ID: | 86 |
Gene Name: | actin like 6A |
Gene Aliases: |
ACTL6, ARPN-BETA, Arp4, BAF53A, INO80K |
Location: |
Chr.3:179562880-179588405 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTL6A | NM_004301.4 | NP_004292.1 | ||
| NM_177989.3 | NP_817126.1 | |||
| NM_178042.3 | NP_829888.1 | |||
| AA256726.1 | ||||
| AB015907.1 | BAA74577.1 | |||
| AB060168.1 | BAB87844.1 | |||
| AB061315.1 | BAB87848.1 | |||
| AF041474.1 | AAC94991.1 | |||
| AK021756.1 | ||||
| AK098691.1 | ||||
| AK223212.1 | BAD96932.1 | |||
| AL136608.1 | CAB66543.1 | |||
| BC000949.2 | AAH00949.1 | |||
| BC001391.2 | AAH01391.1 | |||
| BC036371.1 | AAH36371.1 | |||
| CR533529.1 | CAG38560.1 | |||
| DB041143.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76985230] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map