Assay Details
Target Gene Details
Entrez Gene ID: | 55347 |
Gene Name: | abhydrolase domain containing 10 |
Gene Aliases: |
- |
Location: |
Chr.3:111978876-111993368 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD10 | NM_001272069.1 | NP_001258998.1 | ||
| NM_018394.3 | NP_060864.1 | |||
| NR_073570.1 | ||||
| NR_073571.1 | ||||
| XM_011512960.2 | XP_011511262.1 | |||
| AK002204.1 | BAA92133.1 | |||
| AK300018.1 | ||||
| AL713726.1 | CAD28516.2 | |||
| BC014516.2 | AAH14516.1 | |||
| DB494690.1 | ||||
| DR760607.1 |
More Information
Set Membership: |
|

Genomic Map