Assay Details
Target Gene Details
Entrez Gene ID: | 23231 |
Gene Name: | SEL1L family member 3 |
Gene Aliases: |
Sel-1L3 |
Location: |
Chr.4:25697790-25863595 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEL1L3 | NM_001297592.1 | NP_001284521.1 | ||
| NM_001297594.1 | NP_001284523.1 | |||
| NM_015187.4 | NP_056002.2 | |||
| AB018289.1 | BAA34466.1 | |||
| AK025282.1 | ||||
| AK123293.1 | ||||
| AK289687.1 | ||||
| AK300219.1 | ||||
| BC009945.2 | AAH09945.2 | |||
| BC060867.1 | AAH60867.1 | |||
| CR749817.1 | CAH18677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997544 | Chr.4:25362237 - 25915432 on Build GRCh38 | Gain |
|

Genomic Map