Assay Details
Target Gene Details
Entrez Gene ID: | 8382 |
Gene Name: | NME/NM23 family member 5 |
Gene Aliases: |
NM23-H5, NM23H5, RSPH23 |
Location: |
Chr.5:138115172-138139443 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NME5 | NM_003551.2 | NP_003542.1 | ||
XM_005272099.1 | XP_005272156.1 | |||
XM_006714716.3 | XP_006714779.1 | |||
XM_017009945.1 | XP_016865434.1 | |||
AF067724.1 | AAC64358.1 | |||
AK312181.1 | ||||
BC026182.2 | AAH26182.1 | |||
KU178416.1 | ||||
KU178417.1 | ||||
U90450.1 | AAC69440.1 | |||
Y14992.1 | CAA75226.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
