Assay Details
Target Gene Details
Entrez Gene ID: | 154215 |
Gene Name: | Na+/K+ transporting ATPase interacting 2 |
Gene Aliases: |
FAM77B, NKAIP2, TCBA, TCBA1 |
Location: |
Chr.6:123803840-124825652 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKAIN2 | NM_001040214.2 | NP_001035304.1 | ||
| NM_001300737.1 | NP_001287666.1 | |||
| NM_001300738.1 | NP_001287667.1 | |||
| NM_001300740.1 | NP_001287669.1 | |||
| NM_153355.4 | NP_699186.2 | |||
| XM_011535501.2 | XP_011533803.1 | |||
| XM_011535503.2 | XP_011533805.1 | |||
| XM_017010318.1 | XP_016865807.1 | |||
| XM_017010319.1 | XP_016865808.1 | |||
| AB070452.1 | BAB85477.1 | |||
| AK055326.1 | ||||
| AK308927.1 | ||||
| AK309385.1 | ||||
| AK309407.1 | ||||
| AK309802.1 | ||||
| AK310763.1 | ||||
| AK315886.1 | ||||
| BC035062.2 | AAH35062.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524559 | Chr.6:123189871 - 124008531 on Build GRCh38 | Gain |
|
| nsv1018599 | Chr.6:123187871 - 124008918 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76124961] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map