Assay Details
Target Gene Details
Entrez Gene ID: | 10667 |
Gene Name: | phenylalanyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
COXPD14, FARS1, HSPC320, PheRS |
Location: |
Chr.6:5261001-5771592 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FARS2 | NM_001318872.1 | NP_001305801.1 | ||
NM_006567.4 | NP_006558.1 | |||
XM_005248812.3 | XP_005248869.1 | |||
XM_006714966.2 | XP_006715029.1 | |||
XM_011514247.2 | XP_011512549.1 | |||
XM_011514248.2 | XP_011512550.1 | |||
XM_011514249.1 | XP_011512551.1 | |||
XM_011514251.2 | XP_011512553.1 | |||
XM_017010186.1 | XP_016865675.1 | |||
XM_017010187.1 | XP_016865676.1 | |||
AK223423.1 | BAD97143.1 | |||
AK312454.1 | ||||
BC020239.1 | AAH20239.1 | |||
BC021112.1 | AAH21112.1 | |||
BE792999.1 | ||||
BI761156.1 | ||||
CN292346.1 | ||||
DA134696.1 | ||||
DA516347.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv600873 | Chr.6:5089044 - 5400425 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv5912n100 | Chr.6:5132027 - 5431352 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv1028185 | Chr.6:5247391 - 5339930 on Build GRCh38 | Loss |
![]() ![]() |
nsv600872 | Chr.6:4975079 - 5341957 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
