Assay Details
Target Gene Details
Entrez Gene ID: | 9477 |
Gene Name: | mediator complex subunit 20 |
Gene Aliases: |
PRO0213, TRFP |
Location: |
Chr.6:41905353-41921147 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED20 | NM_001305455.1 | NP_001292384.1 | ||
| NM_001305456.1 | NP_001292385.1 | |||
| NM_001305457.1 | NP_001292386.1 | |||
| NM_004275.4 | NP_004266.2 | |||
| NR_131160.1 | ||||
| AF097725.1 | AAD16169.1 | |||
| AJ420544.1 | ||||
| AK023092.1 | BAB14399.1 | |||
| AK097385.1 | ||||
| AK293412.1 | ||||
| AK300573.1 | ||||
| AK307070.1 | ||||
| BC012618.1 | AAH12618.1 | |||
| BC019866.1 | ||||
| BC032552.1 | AAH32552.1 | |||
| BC040950.1 | AAH40950.1 | |||
| BU597139.1 | ||||
| DA909832.1 |
More Information
Set Membership: |
|

Genomic Map