Assay Details
Target Gene Details
Entrez Gene ID: | 57579 |
Gene Name: | family with sequence similarity 135 member A |
Gene Aliases: |
KIAA1411 |
Location: |
Chr.6:70413404-70561174 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524328 | Chr.6:70252130 - 70849102 on Build GRCh38 | Gain |
|
| nsv1027161 | Chr.6:70396954 - 70435463 on Build GRCh38 | Loss |
|
| nsv1027049 | Chr.6:70251907 - 70851785 on Build GRCh38 | Gain |
|
| nsv1032876 | Chr.6:70338725 - 70435463 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map