Assay Details
Target Gene Details
Entrez Gene ID: | 55227 |
Gene Name: | leucine rich repeat containing 1 |
Gene Aliases: |
LANO, dJ523E19.1 |
Location: |
Chr.6:53794497-53924125 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC1 | NM_018214.4 | NP_060684.4 | ||
| XM_011514726.2 | XP_011513028.1 | |||
| XM_017010997.1 | XP_016866486.1 | |||
| AF332199.1 | AAK69623.1 | |||
| AF359380.1 | AAK72246.1 | |||
| AK001637.1 | BAA91801.1 | |||
| AK021896.1 | BAB13929.1 | |||
| AK292248.1 | ||||
| BC003193.1 | AAH03193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034955 | Chr.6:53582809 - 54066257 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73430242] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map