Assay Details
Target Gene Details
Entrez Gene ID: | 51106 |
Gene Name: | transcription factor B1, mitochondrial |
Gene Aliases: |
CGI-75, CGI75, mtTFB, mtTFB1 |
Location: |
Chr.6:155229990-155314497 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFB1M |
Target Gene Details
Entrez Gene ID: | 26230 |
Gene Name: | T-cell lymphoma invasion and metastasis 2 |
Gene Aliases: |
STEF, TIAM-2 |
Location: |
Chr.6:155090289-155257723 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIAM2 | NM_001010927.2 | NP_001010927.1 | ||
| NM_012454.3 | NP_036586.2 | |||
| AF120323.1 | AAF05900.1 | |||
| AF120324.1 | AAF05901.1 | |||
| AK123859.1 | ||||
| AK125459.1 | BAC86170.1 | |||
| AK299723.1 | ||||
| AK302860.1 | ||||
| AL122086.1 | CAB59259.2 | |||
| BC066979.1 | AAH66979.1 | |||
| BC137275.1 | ||||
| BP199501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526576 | Chr.6:155230087 - 155258266 on Build GRCh38 | Loss |
|
| nsv1025003 | Chr.6:155077125 - 155755140 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map