Assay Details
Target Gene Details
Entrez Gene ID: | 81847 |
Gene Name: | ring finger protein 146 |
Gene Aliases: |
- |
Location: |
Chr.6:127266340-127289893 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF146 | NM_001242844.1 | NP_001229773.1 | ||
| NM_001242845.1 | NP_001229774.1 | |||
| NM_001242846.1 | 3 | 243 | NP_001229775.1 | |
| NM_001242847.1 | NP_001229776.1 | |||
| NM_001242848.1 | NP_001229777.1 | |||
| NM_001242849.1 | NP_001229778.1 | |||
| NM_001242850.1 | NP_001229779.1 | |||
| NM_001242851.1 | NP_001229780.1 | |||
| NM_001242852.1 | NP_001229781.1 | |||
| NM_030963.3 | NP_112225.2 | |||
| XM_006715571.3 | 3 | 911 | XP_006715634.1 | |
| XM_011536161.2 | XP_011534463.1 | |||
| XM_011536162.2 | 4 | 850 | XP_011534464.1 | |
| XM_011536163.2 | 4 | 830 | XP_011534465.1 | |
| XM_011536164.2 | XP_011534466.1 | |||
| XM_017011336.1 | XP_016866825.1 | |||
| XM_017011337.1 | 3 | 3832 | XP_016866826.1 | |
| XM_017011338.1 | 5 | 939 | XP_016866827.1 | |
| XM_017011339.1 | XP_016866828.1 | |||
| XM_017011340.1 | 4 | 1020 | XP_016866829.1 | |
| XM_017011341.1 | XP_016866830.1 | |||
| XM_017011342.1 | 3 | 860 | XP_016866831.1 | |
| XM_017011343.1 | 4 | 969 | XP_016866832.1 | |
| AK027558.1 | BAB55196.1 | |||
| AL136829.1 | CAB66763.1 | |||
| BG708628.1 | ||||
| BI669435.1 | ||||
| BI771613.1 | ||||
| CN388414.1 | ||||
| DA043259.1 | 3 | 198 | ||
| DA145911.1 | ||||
| DA639989.1 | ||||
| DA707096.1 | ||||
| DA826982.1 | ||||
| DB267873.1 |
More Information
Set Membership: |
|

Genomic Map