Assay Details
Target Gene Details
Entrez Gene ID: | 23493 |
Gene Name: | hes related family bHLH transcription factor with YRPW motif 2 |
Gene Aliases: |
CHF1, GRIDLOCK, GRL, HERP1, HESR2, HRT2, bHLHb32 |
Location: |
Chr.6:125747592-125762243 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HEY2 | NM_012259.2 | NP_036391.1 | ||
XM_017010627.1 | XP_016866116.1 | |||
XM_017010628.1 | XP_016866117.1 | |||
XM_017010629.1 | XP_016866118.1 | |||
AB044755.1 | BAA96781.1 | |||
AF173901.1 | AAF20173.1 | |||
AF232238.1 | AAF37296.1 | |||
AF237949.1 | AAF44781.1 | |||
AF311884.1 | AAG31157.1 | |||
AJ249545.1 | CAB56839.1 | |||
AK289943.1 | ||||
AK303890.1 | ||||
BC007707.2 | AAH07707.1 | |||
BT020067.1 | AAV38870.1 | |||
BT020068.1 | AAV38871.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
