Assay Details
Target Gene Details
Entrez Gene ID: | 54898 |
Gene Name: | ELOVL fatty acid elongase 2 |
Gene Aliases: |
SSC2 |
Location: |
Chr.6:10980759-11044391 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL2 | NM_017770.3 | NP_060240.3 | ||
| XM_011514716.2 | XP_011513018.1 | |||
| XM_011514717.2 | XP_011513019.1 | |||
| AK000341.1 | BAA91096.1 | |||
| BC050278.1 | AAH50278.2 | |||
| BC060809.1 | AAH60809.1 |
More Information
Set Membership: |
|

Genomic Map