Assay Details
Target Gene Details
Entrez Gene ID: | 167681 |
Gene Name: | protease, serine 35 |
Gene Aliases: |
C6orf158, dJ223E3.1 |
Location: |
Chr.6:83512475-83525704 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS35 | NM_001170423.1 | NP_001163894.1 | ||
| NM_153362.2 | NP_699193.2 | |||
| AK291928.1 | ||||
| AY358661.1 | AAQ89024.1 | |||
| BC037170.1 | AAH37170.1 | |||
| DA251169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3571277 | Chr.6:83434599 - 83518745 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112550597] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map