Assay Details
Target Gene Details
Entrez Gene ID: | 285848 |
Gene Name: | patatin like phospholipase domain containing 1 |
Gene Aliases: |
ARCI10, dJ50J22.1 |
Location: |
Chr.6:36242523-36313955 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNPLA1 | XM_011514520.2 | 1 | 284 | XP_011512822.1 |
| XM_017010778.1 | 1 | 284 | XP_016866267.1 |
More Information
Set Membership: |
|

Genomic Map