Assay Details
Target Gene Details
Entrez Gene ID: | 89845 |
Gene Name: | ATP binding cassette subfamily C member 10 |
Gene Aliases: |
EST182763, MRP7, SIMRP7 |
Location: |
Chr.6:43427538-43451994 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC10 | NM_001198934.1 | NP_001185863.1 | ||
| XM_011514983.2 | XP_011513285.1 | |||
| XM_017011446.1 | XP_016866935.1 | |||
| XM_017011447.1 | XP_016866936.1 | |||
| XM_017011452.1 | XP_016866941.1 | |||
| AK000002.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv603000 | Chr.6:43397622 - 43524840 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map