Assay Details
Target Gene Details
Entrez Gene ID: | 285755 |
Gene Name: | peptidylprolyl isomerase like 6 |
Gene Aliases: |
PPIase, RSPH12, bA425D10.6, dJ919F19.1 |
Location: |
Chr.6:109390215-109441171 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIL6 | NM_001111298.2 | NP_001104768.2 | ||
| NM_001286360.1 | NP_001273289.1 | |||
| NM_001286361.1 | NP_001273290.1 | |||
| NM_173672.4 | NP_775943.1 | |||
| NR_104429.1 | ||||
| XM_011535765.2 | XP_011534067.1 | |||
| XM_017010774.1 | XP_016866263.1 | |||
| AK293674.1 | ||||
| AK315790.1 | ||||
| BC038716.1 | AAH38716.1 | |||
| BI824055.1 | ||||
| DQ363562.1 | ||||
| DQ423529.1 |
More Information
Set Membership: |
|

Genomic Map