Assay Details
Target Gene Details
Entrez Gene ID: | 8528 |
Gene Name: | D-aspartate oxidase |
Gene Aliases: |
DASOX, DDO-1, DDO-2 |
Location: |
Chr.6:110392180-110415550 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDO | NM_003649.2 | NP_003640.2 | ||
| NM_004032.2 | NP_004023.2 | |||
| AB496420.1 | ||||
| AK293029.1 | ||||
| AX786391.1 | CAE11960.1 | |||
| BC032786.1 | AAH32786.3 | |||
| D89858.1 | BAA14031.1 |
More Information
Set Membership: |
|

Genomic Map