Assay Details
Target Gene Details
Entrez Gene ID: | 57480 |
Gene Name: | pleckstrin homology and RhoGEF domain containing G1 |
Gene Aliases: |
ARHGEF41 |
Location: |
Chr.6:150599863-150843665 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHG1 | NM_001029884.1 | NP_001025055.1 | ||
| XM_005267065.3 | XP_005267122.1 | |||
| AK293288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3611236 | Chr.6:150598977 - 150602526 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117813850] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map