Assay Details
Target Gene Details
Entrez Gene ID: | 135138 |
Gene Name: | PARK2 coregulated |
Gene Aliases: |
GLUP, HAK005771, PARK2CRG |
Location: |
Chr.6:162726870-163315492 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PACRG | NM_001080378.1 | NP_001073847.1 | ||
| NM_001080379.1 | NP_001073848.1 | |||
| NM_152410.2 | NP_689623.2 | |||
| XM_011535461.2 | XP_011533763.1 | |||
| XM_011535462.2 | XP_011533764.1 | |||
| AF546872.1 | AAN37911.1 | |||
| AK057286.1 | BAB71410.1 | |||
| BC030642.2 | AAH30642.1 | |||
| BC044227.1 | AAH44227.1 |
Target Gene Details
Entrez Gene ID: | 729658 |
Gene Name: | PACRG antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.6:163181645-163192002 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PACRG-AS3 | NR_122120.1 | 4 | 1744 | |
| AK296276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026490 | Chr.6:162712401 - 163284676 on Build GRCh38 | Gain |
|
| esv2762628 | Chr.6:163151116 - 163199871 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map