Assay Details
Target Gene Details
Entrez Gene ID: | 2911 |
Gene Name: | glutamate metabotropic receptor 1 |
Gene Aliases: |
GPRC1A, MGLU1, MGLUR1, PPP1R85, SCAR13 |
Location: |
Chr.6:146027634-146437598 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM1 | NM_001278064.1 | NP_001264993.1 | ||
| NM_001278065.1 | NP_001264994.1 | |||
| NM_001278066.1 | NP_001264995.1 | |||
| NM_001278067.1 | NP_001264996.1 | |||
| XM_011535782.1 | XP_011534084.1 | |||
| XM_017010783.1 | XP_016866272.1 | |||
| XM_017010784.1 | XP_016866273.1 | |||
| XM_017010785.1 | XP_016866274.1 | |||
| XM_017010786.1 | XP_016866275.1 | |||
| XM_017010787.1 | XP_016866276.1 | |||
| AB208837.1 | BAD92074.1 | |||
| AK308550.1 | ||||
| BC136280.1 | ||||
| L76627.1 | AAB05337.1 | |||
| L76631.1 | AAB05338.1 | |||
| U31215.1 | AAA87843.1 | |||
| U31216.1 | AAA87844.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604839 | Chr.6:145984674 - 146507786 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map