Assay Details
Target Gene Details
Entrez Gene ID: | 285796 |
Gene Name: | PACRG antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:163309985-163324473 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PACRG-AS1 | NR_028390.1 | |||
| AK094729.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509165 | Chr.6:163295067 - 163371517 on Build GRCh38 | Insertion |
|
| nsv605188 | Chr.6:163276407 - 163351064 on Build GRCh38 | Loss |
|

Genomic Map