Assay Details
Target Gene Details
Entrez Gene ID: | 4199 |
Gene Name: | malic enzyme 1 |
Gene Aliases: |
HUMNDME, MES |
Location: |
Chr.6:83210389-83431219 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ME1 | NM_002395.5 | NP_002386.1 | ||
XM_011535836.2 | XP_011534138.1 | |||
AK223417.1 | BAD97137.1 | |||
AK289783.1 | ||||
AK301875.1 | ||||
AK302777.1 | ||||
BC017403.1 | AAH17403.1 | |||
BC025246.1 | AAH25246.1 | |||
L34035.1 | AAB01380.1 | |||
U43944.1 | AAC50613.1 | |||
X77244.1 | CAA54460.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
