Assay Details
Target Gene Details
Entrez Gene ID: | 8875 |
Gene Name: | vanin 2 |
Gene Aliases: |
FOAP-4, GPI-80 |
Location: |
Chr.6:132743503-132763489 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VNN2 | NM_001242350.2 | NP_001229279.1 | ||
| NM_004665.4 | NP_004656.2 | |||
| NM_078488.2 | NP_511043.1 | |||
| NR_034173.2 | ||||
| NR_034174.2 | ||||
| NR_110143.1 | ||||
| NR_110144.1 | ||||
| NR_110145.1 | ||||
| NR_110146.1 | ||||
| XM_006715593.3 | XP_006715656.1 | |||
| XM_011536231.1 | XP_011534533.1 | |||
| XM_017011407.1 | XP_016866896.1 | |||
| XM_017011408.1 | XP_016866897.1 | |||
| XM_017011409.1 | XP_016866898.1 | |||
| AB026705.1 | BAB61019.1 | |||
| AB435062.1 | ||||
| AB435063.1 | ||||
| AB435064.1 | ||||
| AB435065.1 | ||||
| AJ132100.1 | CAA10569.1 | |||
| AK290908.1 | ||||
| AK292125.1 | ||||
| AK298719.1 | ||||
| BC126145.1 | ||||
| BC126147.1 | ||||
| D89974.1 | BAA82525.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3610843 | Chr.6:132752877 - 132755889 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map