Assay Details
Target Gene Details
Entrez Gene ID: | 9587 |
Gene Name: | MAD2L1 binding protein |
Gene Aliases: |
CMT2 |
Location: |
Chr.6:43629542-43640952 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAD2L1BP | NM_001003690.1 | NP_001003690.1 | ||
| AL537692.3 | ||||
| BE297042.1 | ||||
| BU845738.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5281 | Chr.6:43611090 - 43655905 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map