Assay Details
Target Gene Details
Entrez Gene ID: | 93643 |
Gene Name: | tight junction associated protein 1 |
Gene Aliases: |
PILT, TJP4 |
Location: |
Chr.6:43477523-43506557 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv603000 | Chr.6:43397622 - 43524840 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map