Assay Details
Target Gene Details
Entrez Gene ID: | 7172 |
Gene Name: | thiopurine S-methyltransferase |
Gene Aliases: |
TPMTD |
Location: |
Chr.6:18128311-18155165 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPMT | NM_000367.3 | NP_000358.1 | ||
XM_011514839.2 | XP_011513141.1 | |||
XM_017011241.1 | XP_016866730.1 | |||
AF021876.1 | AAB80746.1 | |||
AF021877.1 | AAB80747.1 | |||
AK310420.1 | ||||
BC005339.1 | AAH05339.1 | |||
BC009596.1 | AAH09596.1 | |||
BI598325.1 | ||||
S62904.1 | AAB27277.1 | |||
U12387.1 | AAC50130.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1034755 | Chr.6:18064320 - 18173781 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv601050 | Chr.6:18134947 - 18221240 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
