Assay Details
Target Gene Details
Entrez Gene ID: | 3096 |
Gene Name: | human immunodeficiency virus type I enhancer binding protein 1 |
Gene Aliases: |
CIRIP, CRYBP1, GAAP, MBP-1, PRDII-BF1, Schnurri-1, ZAS1, ZNF40, ZNF40A |
Location: |
Chr.6:12002096-12212034 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIVEP1 | XM_017010801.1 | XP_016866290.1 |
More Information
Set Membership: |
|

Genomic Map