Assay Details
Target Gene Details
Entrez Gene ID: | 285740 |
Gene Name: | PHACTR2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:143554115-143569339 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHACTR2-AS1 | NR_027113.2 | |||
| NR_027114.2 | ||||
| NR_110148.1 | ||||
| NR_110149.1 | ||||
| AK093151.1 | ||||
| AK098520.1 | ||||
| HY000603.1 | ||||
| HY207533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3611097 | Chr.6:143554635 - 143556416 on Build GRCh38 | Loss |
|

Genomic Map