Assay Details
Target Gene Details
Entrez Gene ID: | 51362 |
Gene Name: | cell division cycle 40 |
Gene Aliases: |
EHB3, PRP17, PRPF17 |
Location: |
Chr.6:110180421-110232220 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC40 | NM_015891.2 | NP_056975.1 | ||
| AB208903.1 | BAD92140.1 | |||
| AF015044.1 | AAD01552.1 | |||
| AF038392.1 | AAC39730.1 | |||
| AF061241.1 | AAC25166.1 | |||
| AK222721.1 | BAD96441.1 | |||
| AK295055.1 | ||||
| AK314601.1 | ||||
| BC111061.1 | AAI11062.1 | |||
| BC117145.1 | ||||
| BC126114.1 |
More Information
Set Membership: |
|

Genomic Map