Assay Details
Target Gene Details
Entrez Gene ID: | 100506409 |
Gene Name: | ELOVL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:11043758-11079144 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL2-AS1 | NR_038962.1 | |||
| NR_038963.1 | ||||
| NR_038964.1 | ||||
| BI828717.1 | ||||
| DN918479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5198 | Chr.6:11042676 - 11074421 on Build GRCh38 | Insertion |
|

Genomic Map