Assay Details
Target Gene Details
Entrez Gene ID: | 5110 |
Gene Name: | protein-L-isoaspartate (D-aspartate) O-methyltransferase |
Gene Aliases: |
PIMT |
Location: |
Chr.6:149749695-149811421 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCMT1 | NM_001252049.1 | NP_001238978.1 | ||
NM_001252050.1 | NP_001238979.1 | |||
NM_001252051.1 | NP_001238980.1 | |||
NM_001252052.1 | NP_001238981.1 | |||
NM_001252053.1 | NP_001238982.1 | |||
NM_005389.2 | NP_005380.2 | |||
AK098739.1 | ||||
AK129778.1 | ||||
AK289724.1 | ||||
AK304532.1 | ||||
AL539512.3 | ||||
BC007501.2 | AAH07501.1 | |||
BC008748.2 | AAH08748.1 | |||
BE262147.1 | ||||
BX438393.2 | ||||
CX786396.1 | ||||
D13892.1 | BAA02991.1 | |||
D25545.1 | BAA05028.1 | |||
D25546.1 | BAA05029.1 | |||
D25547.1 | BAA05030.1 | |||
DB478200.1 | ||||
DC339361.1 | ||||
DC356391.1 | ||||
DC421281.1 | ||||
M93008.1 | AAA90934.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2732861 | Chr.6:149681836 - 149764933 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
