Assay Details
Target Gene Details
Entrez Gene ID: | 3066 |
Gene Name: | histone deacetylase 2 |
Gene Aliases: |
HD2, RPD3, YAF1 |
Location: |
Chr.6:113936156-113971195 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC2 | NM_001527.3 | NP_001518.3 | ||
| NR_033441.1 | ||||
| NR_073443.1 | ||||
| XM_011535788.1 | XP_011534090.1 | |||
| XM_017010799.1 | XP_016866288.1 | |||
| AB209190.1 | 3 | 2055 | BAD92427.1 | |
| AK092156.1 | ||||
| AK097376.1 | ||||
| AK296856.1 | ||||
| BC031055.1 | AAH31055.2 | |||
| U31814.1 | AAC50814.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79278914] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map