Assay Details
Target Gene Details
| Entrez Gene ID: | 158038 | 
| Gene Name: | leucine rich repeat and Ig domain containing 2 | 
| Gene Aliases: | LERN3, LRRN6C | 
| Location: | Chr.9:27937617-29213981 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 14 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| LINGO2 | NM_001258282.1 | NP_001245211.1 | ||
| NM_152570.2 | NP_689783.1 | |||
| XM_011517719.1 | XP_011516021.1 | |||
| XM_011517724.2 | XP_011516026.1 | |||
| XM_011517728.2 | XP_011516030.1 | |||
| XM_017014303.1 | XP_016869792.1 | |||
| XM_017014304.1 | XP_016869793.1 | |||
| XM_017014305.1 | XP_016869794.1 | |||
| XM_017014306.1 | XP_016869795.1 | |||
| XM_017014307.1 | XP_016869796.1 | |||
| AK056372.1 | BAB71167.1 | |||
| AK290972.1 | ||||
| AY358114.1 | AAQ88481.1 | |||
| BC137514.1 | ||||
| BC144676.1 | ||||
| DN991981.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv613950 | Chr.9:27746787 - 28342181 on Build GRCh38 | Gain |  LINGO2  LOC646700 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top