Assay Details
Target Gene Details
Entrez Gene ID: | 105376204 |
Gene Name: | uncharacterized LOC105376204 |
Gene Aliases: |
- |
Location: |
Chr.9:106852964-106863015 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105376204 | XM_017015409.1 | 1 | 8869 | XP_016870898.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79080672] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map