Assay Details
Target Gene Details
Entrez Gene ID: | 26086 |
Gene Name: | G-protein signaling modulator 1 |
Gene Aliases: |
AGS3 |
Location: |
Chr.9:136327476-136359605 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPSM1 | NM_001145638.2 | NP_001139110.2 | ||
| NM_015597.5 | NP_056412.5 | |||
| XM_011518498.2 | XP_011516800.1 | |||
| XM_017014598.1 | XP_016870087.1 | |||
| AY173053.1 | AAO17260.1 | |||
| BC009979.1 | AAH09979.1 | |||
| BC017353.1 | AAH17353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046074 | Chr.9:136228197 - 136393924 on Build GRCh38 | Gain |
|
| nsv825173 | Chr.9:135892081 - 136495763 on Build GRCh38 | Gain |
|
| nsv951202 | Chr.9:135770355 - 136558248 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map